ENA424046234 Enamine

1-(propan-2-yl)-1H-imidazole

  • MDL number MFCD00234239
  • Cas Number : 4532-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79714

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79714
Availability details for
Quantity
  • Availability
  • Add to Cart