ENA424046235 Enamine

tert-butyl N-[4-(trifluoromethyl)phenyl]carbamate

  • MDL number MFCD09859792
  • Cas Number : 141940-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83010

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83010
Availability details for
Quantity
  • Availability
  • Add to Cart