ENA424046248 Enamine

2-(cyclopropylmethyl)-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD23726250
  • Cas Number : 1529418-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92189
Availability details for
Quantity
  • Availability
  • Add to Cart