ENA424046254 Enamine

1-ethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD12923515
  • Cas Number : 400715-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-96447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96447
Availability details for
Quantity
  • Availability
  • Add to Cart