ENA428072374 Enamine

methyl 2-[5-(trifluoromethyl)-1,3,4-oxadiazol-2-yl]acetate

  • MDL number MFCD28023705
  • Cas Number : 1803600-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158464
Availability details for
Quantity
  • Availability
  • Add to Cart