ENA428072376 Enamine

1-[2-(trifluoromethyl)-5H,6H,7H,8H-imidazo[1,2-a]pyridin-6-yl]methanamine

  • MDL number MFCD28023698
  • Cas Number : 1694748-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158522
Availability details for
Quantity
  • Availability
  • Add to Cart