ENA428072379 Enamine

5-(1-acetylpiperidin-2-yl)-1-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD28023701
  • Cas Number : 1788685-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-158627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158627
Availability details for
Quantity
  • Availability
  • Add to Cart