ENA428072380 Enamine

N,1-dimethyl-5-(piperidin-2-yl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD28023706
  • Cas Number : 1803590-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-158631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158631
Availability details for
Quantity
  • Availability
  • Add to Cart