ENA428072384 Enamine

tert-butyl 4-{[(4-aminopyrimidin-2-yl)methyl]amino}azepane-1-carboxylate

  • MDL number MFCD28023722
  • Cas Number : 1803611-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-158988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158988
Availability details for
Quantity
  • Availability
  • Add to Cart