ENA428072391 Enamine

1-(oxan-3-yl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD28023725
  • Cas Number : 1803598-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-159667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159667
Availability details for
Quantity
  • Availability
  • Add to Cart