ENA428072394 Enamine

N'-hydroxy-2-(oxan-2-yloxy)propanimidamide

  • Cas Number : 1807888-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160422
Availability details for
Quantity
  • Availability
  • Add to Cart