ENA428072397 Enamine

2,2-difluoro-2-(1,3-thiazol-2-yl)ethan-1-ol

  • MDL number MFCD28023709
  • Cas Number : 1803604-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-170933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170933
Availability details for
Quantity
  • Availability
  • Add to Cart