ENA428072398 Enamine

4-{[(tert-butoxy)carbonyl]amino}-3-methoxybutanoic acid

  • MDL number MFCD28023728
  • Cas Number : 1697729-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-170969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170969
Availability details for
Quantity
  • Availability
  • Add to Cart