ENA428072399 Enamine

2-(1,1-diethoxyethyl)-1,3,4-oxadiazole

  • MDL number MFCD28023710
  • Cas Number : 1803584-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171275
Availability details for
Quantity
  • Availability
  • Add to Cart