ENA428072406 Enamine

3-phenylazetidin-3-ol; trifluoroacetic acid

  • MDL number MFCD28063899
  • Cas Number : 958297-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-172443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172443
Availability details for
Quantity
  • Availability
  • Add to Cart