ENA428072410 Enamine

1-(carbamoylmethyl)-2-oxocyclohexane-1-carboxylic acid

  • MDL number MFCD28023733
  • Cas Number : 1803599-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-172923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-172923
Availability details for
Quantity
  • Availability
  • Add to Cart