ENA428072412 Enamine

methyl 2-[(3-nitropyridin-4-yl)oxy]acetate

  • MDL number MFCD28023712
  • Cas Number : 592552-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173580
Availability details for
Quantity
  • Availability
  • Add to Cart