ENA428072421 Enamine

tert-butyl 3-[N'-(2-methylpropanoyl)hydrazinecarbonyl]morpholine-4-carboxylate

  • MDL number MFCD28023755
  • Cas Number : 1803561-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-174385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-174385
Availability details for
Quantity
  • Availability
  • Add to Cart