ENA428072426 Enamine

5-(1-chloroethyl)-1-cyclopropyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD28023757
  • Cas Number : 1700118-39-3
  • Molecular Weight : 172.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-174585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-174585
Availability details for
Quantity
  • Availability
  • Add to Cart