ENA428072431 Enamine

3-methyl-N-{[(propan-2-yl)carbamoyl]amino}oxolane-2-carboxamide

  • MDL number MFCD28023717
  • Cas Number : 1803587-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-174895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-174895
Availability details for
Quantity
  • Availability
  • Add to Cart