ENA428072437 Enamine

methyl 5-(1-hydroxyethyl)furan-3-carboxylate

  • MDL number MFCD28064217
  • Cas Number : 1803605-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175410
Availability details for
Quantity
  • Availability
  • Add to Cart