ENA428072440 Enamine

[3-(4-{[(tert-butyldimethylsilyl)oxy]methyl}phenyl)prop-1-yn-1-yl]trimethylsilane

  • MDL number MFCD28023763
  • Cas Number : 1803601-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-175533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-175533
Availability details for
Quantity
  • Availability
  • Add to Cart