ENA428072444 Enamine

1-cyclopentylpiperidine-2-carboxylic acid hydrochloride

  • MDL number MFCD28023777
  • Cas Number : 1786116-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-175748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-175748
Availability details for
Quantity
  • Availability
  • Add to Cart