ENA428072452 Enamine

5-(chloromethyl)-2-methylquinolin-8-ol hydrochloride

  • MDL number MFCD28023739
  • Cas Number : 103862-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176249
Availability details for
Quantity
  • Availability
  • Add to Cart