ENA428072459 Enamine

sodium 4-hydroxypentanoate

  • MDL number MFCD28023743
  • Cas Number : 56279-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176456
Availability details for
Quantity
  • Availability
  • Add to Cart