ENA428072460 Enamine

6-chloro-3-[(hydroxyimino)methyl]pyridin-2-amine

  • Cas Number : 1062369-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176528
Availability details for
Quantity
  • Availability
  • Add to Cart