ENA428072461 Enamine

N-methoxy-2-(1-methoxycyclobutyl)-N-methylacetamide

  • MDL number MFCD28064224
  • Cas Number : 1698246-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176538
Availability details for
Quantity
  • Availability
  • Add to Cart