ENA428072463 Enamine

methyl 2-(methoxyamino)propanoate hydrochloride

  • MDL number MFCD28023789

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176556
Availability details for
Quantity
  • Availability
  • Add to Cart