ENA428072466 Enamine

1-{3-[(difluoromethyl)sulfanyl]phenyl}methanamine hydrochloride

  • MDL number MFCD28023597
  • Cas Number : 1803584-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176633
Availability details for
Quantity
  • Availability
  • Add to Cart