ENA428072469 Enamine

6-(bromomethyl)-5-oxaspiro[2.5]octan-4-one

  • MDL number MFCD28064246
  • Cas Number : 1803587-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176653
Availability details for
Quantity
  • Availability
  • Add to Cart