ENA428072470 Enamine

3-ethoxy-2-methanesulfonylpent-2-enenitrile

  • Cas Number : 1704400-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176657
Availability details for
Quantity
  • Availability
  • Add to Cart