ENA428072471 Enamine

1-[4-(chloromethyl)-3-methylphenyl]-1H-imidazole hydrochloride

  • MDL number MFCD28023599
  • Cas Number : 1803585-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176701
Availability details for
Quantity
  • Availability
  • Add to Cart