ENA428072473 Enamine

2,6-dimethyl-1lambda6-thiomorpholine-1,1-dione hydrochloride

  • MDL number MFCD28023748
  • Cas Number : 1803601-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176745
Availability details for
Quantity
  • Availability
  • Add to Cart