ENA428072485 Enamine

2-(3-methyl-1,2,4-oxadiazol-5-yl)-1-phenylethan-1-amine hydrochloride

  • MDL number MFCD28023613
  • Cas Number : 1803590-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177018
Availability details for
Quantity
  • Availability
  • Add to Cart