ENA428072486 Enamine

5-chloro-2-methylquinolin-6-ol hydrobromide

  • MDL number MFCD28023614
  • Cas Number : 1803584-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177024
Availability details for
Quantity
  • Availability
  • Add to Cart