ENA428072488 Enamine

(2S)-2-(4-chlorophenoxy)propan-1-ol

  • MDL number MFCD28064251
  • Cas Number : 87810-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177042
Availability details for
Quantity
  • Availability
  • Add to Cart