ENA428072491 Enamine

N5-(azepan-4-yl)-N2,N2-dimethylpyridine-2,5-diamine trihydrochloride

  • MDL number MFCD28023618
  • Cas Number : 1803581-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177071
Availability details for
Quantity
  • Availability
  • Add to Cart