ENA428072495 Enamine

5-chloro-6-methoxy-2-methylquinoline hydrochloride

  • MDL number MFCD28023623
  • Cas Number : 1803607-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177164
Availability details for
Quantity
  • Availability
  • Add to Cart