ENA428072498 Enamine

3-(dimethylamino)-3-phenylpropanoic acid hydrochloride

  • MDL number MFCD28023624
  • Cas Number : 85608-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177249
Availability details for
Quantity
  • Availability
  • Add to Cart