ENA428072502 Enamine

N-(2,2-dichloro-1-cyanoeth-1-en-1-yl)-6-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)hexanamide

  • MDL number MFCD28064259
  • Cas Number : 1652554-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177290
Availability details for
Quantity
  • Availability
  • Add to Cart