ENA428072503 Enamine

6-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-N-(1,2,2,2-tetrachloroethyl)hexanamide

  • MDL number MFCD28064260
  • Cas Number : 1443664-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177294
Availability details for
Quantity
  • Availability
  • Add to Cart