ENA428072504 Enamine

7-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-N-(2,2,2-trichloro-1-hydroxyethyl)heptanamide

  • MDL number MFCD28064261
  • Cas Number : 1803571-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177296
Availability details for
Quantity
  • Availability
  • Add to Cart