ENA428072505 Enamine

7-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)heptanamide

  • MDL number MFCD28064262
  • Cas Number : 1803595-08-5
  • Molecular Weight : 274.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-177298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177298
Availability details for
Quantity
  • Availability
  • Add to Cart