ENA428072507 Enamine

1-(3-chlorophenyl)-5-(propan-2-yl)-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD28023627
  • Cas Number : 1803567-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177376
Availability details for
Quantity
  • Availability
  • Add to Cart