ENA428072514 Enamine

6-chloro-5-fluoropyridine-3-sulfonamide

  • MDL number MFCD28064234
  • Cas Number : 1803571-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177528
Availability details for
Quantity
  • Availability
  • Add to Cart