ENA428072525 Enamine

1-(benzylsulfanyl)-5-bromo-4-fluoro-2-nitrobenzene

  • MDL number MFCD28064265
  • Cas Number : 1803586-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177784
Availability details for
Quantity
  • Availability
  • Add to Cart