ENA428072527 Enamine

6-fluoroquinoline-3-sulfonyl chloride

  • MDL number MFCD28064266
  • Cas Number : 1803589-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177816
Availability details for
Quantity
  • Availability
  • Add to Cart