ENA428072530 Enamine

1-(3,5,6-trifluoropyridin-2-yl)piperazine

  • MDL number MFCD28064269
  • Cas Number : 1226516-20-6
  • Molecular Weight : 217.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-177924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177924
Availability details for
Quantity
  • Availability
  • Add to Cart