ENA428072532 Enamine

methyl 3-amino-4,6-dibromo-2-methylbenzoate

  • MDL number MFCD25292821
  • Cas Number : 119916-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177943
Availability details for
Quantity
  • Availability
  • Add to Cart