ENA428072534 Enamine

1-(1,3-dioxolan-2-yl)propan-2-amine; oxalic acid

  • MDL number MFCD28023650
  • Cas Number : 1803586-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HOOCCOOH
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177980
Availability details for
Quantity
  • Availability
  • Add to Cart